Learn More
Waters Corp Waters Corporation OBD Prep Column, Phenyl, 5 μm, 130 Å, 30 mm X 75 mm
Catalog No. 50781649 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Phenyl columns offer complementary selectivity compared to C18 stationary phases, particularly for polyaromatic compounds. This product features the most chemically stable commercially available Phenyl sorbent on the market, providing ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Complementary selectivity to C18 stationary phases
  • Most chemically stable commercially available Phenyl sorbent
  • Ultra-low column bleed
  • Long column lifetimes
  • Excellent peak shape
Catalog No. 50-781-649 Supplier Waters Corp Supplier No. 186003278
Add to Cart
Add to Cart
Request a Quote
XBridge BEH Phenyl columns offer complementary selectivity compared to C18 stationary phases, particularly for polyaromatic compounds. This product features the most chemically stable commercially available Phenyl sorbent on the market, providing ultra-low column bleed, long column lifetimes, and excellent peak shape.
  • Complementary selectivity to C18 stationary phases
  • Most chemically stable commercially available Phenyl sorbent
  • Ultra-low column bleed
  • Long column lifetimes
  • Excellent peak shape