Learn More
Waters Corp Waters XBridge BEH Phenyl Preparative Column, 5 μm, 19 mm x 50 mm
Catalog No. 50781708 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These preparative columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. Featuring a chemically stable Phenyl sorbent, this product ensures ultra-low column bleed, extended column lifetimes, and excellent peak shape. Optimum Bed Density (OBD) Technology enables direct scalability from analytical to preparative chromatography, while Ethylene-Bridged Hybrid (BEH) Technology offers universal compound suitability and stability across a wide pH range and high temperatures.
  • Complementary selectivity for polyaromatic compounds
  • Chemically stable Phenyl sorbent for ultra-low column bleed
  • Extended column lifetimes and excellent peak shape
  • OBD Technology provides uniform packed bed density and scalability
  • BEH Technology offers universal compound suitability
  • Stable across a wide pH range and at high temperatures
Catalog No. 50-781-708 Supplier Waters Corp Supplier No. 186003356
These preparative columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. Featuring a chemically stable Phenyl sorbent, this product ensures ultra-low column bleed, extended column lifetimes, and excellent peak shape. Optimum Bed Density (OBD) Technology enables direct scalability from analytical to preparative chromatography, while Ethylene-Bridged Hybrid (BEH) Technology offers universal compound suitability and stability across a wide pH range and high temperatures.
  • Complementary selectivity for polyaromatic compounds
  • Chemically stable Phenyl sorbent for ultra-low column bleed
  • Extended column lifetimes and excellent peak shape
  • OBD Technology provides uniform packed bed density and scalability
  • BEH Technology offers universal compound suitability
  • Stable across a wide pH range and at high temperatures