Learn More
Waters Corp Waters Prep Guard Cartridge, 130Å, 10 μm, 19 mm × 10 mm
Supplier:  Waters Corp 186003991
Encompass_Preferred
This Prep Guard Cartridge complements BEH C18 and C8 phases, serving as a vital tool for HPLC method development. Its embedded carbamate group within the bonded phase ligand offers an alternate selectivity, particularly beneficial for phenolic compounds, when compared to traditional straight-chain alkyl columns.
  • Complements BEH C18 and C8 phases for enhanced method development.
  • Embedded carbamate group provides alternate selectivity.
  • Especially effective for analyzing phenolic compounds.
  • Designed for preparative HPLC systems.
Catalog No. 50-782-113