Learn More
Waters Corp Preparative Column, BEH Shield RP18, 50 x 100 mm, 10 μm
Catalog No. 50782121 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These preparative columns are designed to enhance productivity in preparative high-performance liquid chromatography (HPLC) by offering higher recoveries and extended column lifetimes. The Optimum Bed Density (OBD™) design ensures predictable, uniform density profiles within the column, contributing to exceptional resistance against mechanical chromatographic bed failure and consistent column-to-column performance.
  • Minimizes voids and prevents over-compression during the packing process.
  • Facilitates fast, efficient, lab-scale separations.
  • Enables direct scaling from UPLC, UHPLC, or HPLC screening to lab-scale purification.
  • Utilizes robust chromatographic particles specifically engineered for purification applications.
  • BEH C18 material offers excellent ruggedness and longevity across a wide pH range.
Catalog No. 50-782-121 Supplier Waters Corp Supplier No. 186003999
Add to Cart
Add to Cart
Request a Quote
These preparative columns are designed to enhance productivity in preparative high-performance liquid chromatography (HPLC) by offering higher recoveries and extended column lifetimes. The Optimum Bed Density (OBD™) design ensures predictable, uniform density profiles within the column, contributing to exceptional resistance against mechanical chromatographic bed failure and consistent column-to-column performance.
  • Minimizes voids and prevents over-compression during the packing process.
  • Facilitates fast, efficient, lab-scale separations.
  • Enables direct scaling from UPLC, UHPLC, or HPLC screening to lab-scale purification.
  • Utilizes robust chromatographic particles specifically engineered for purification applications.
  • BEH C18 material offers excellent ruggedness and longevity across a wide pH range.