Learn More
Waters Corp Waters OBD Prep Column, Shield RP18, 130Å, 5 μm, 50 mm × 100 mm
Catalog No. 50782074 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Shield RP18 OBD Prep Columns provide alternate selectivity and improved peak shape for basic compounds. With OBD Technology, they ensure uniform packed beds, bridging the performance gap in preparative LC. They offer direct scalability from analytical to preparative LC with excellent lifetime and consistent performance.
  • Complements BEH C18 and C8 phases for alternate selectivity.
  • Provides unique selectivity and improved peak shape for basic compounds.
  • Ensures uniform, optimally packed column beds.
  • Facilitates uncompromised scale-up from UPLC to preparative LC.
  • Offers extended column lifetimes and maximized recoveries.
  • Reduces method development risk.
  • Stable across a wide pH range (2–11) and at high temperatures.
  • Supports high-mass loading applications with superior peak shapes.
Catalog No. 50-782-074 Supplier Waters Corp Supplier No. 186003939
Add to Cart
Add to Cart
Request a Quote
XBridge BEH Shield RP18 OBD Prep Columns provide alternate selectivity and improved peak shape for basic compounds. With OBD Technology, they ensure uniform packed beds, bridging the performance gap in preparative LC. They offer direct scalability from analytical to preparative LC with excellent lifetime and consistent performance.
  • Complements BEH C18 and C8 phases for alternate selectivity.
  • Provides unique selectivity and improved peak shape for basic compounds.
  • Ensures uniform, optimally packed column beds.
  • Facilitates uncompromised scale-up from UPLC to preparative LC.
  • Offers extended column lifetimes and maximized recoveries.
  • Reduces method development risk.
  • Stable across a wide pH range (2–11) and at high temperatures.
  • Supports high-mass loading applications with superior peak shapes.