Learn More
Waters Corp WATERS CORPORATION XBridge BEH Shield RP18 OBD Prep Column, 130Å, 5 μm, 50 mm × 50 mm
Catalog No. 50782070 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH Shield RP18 columns are designed to complement XBridge BEH C18 and C8 phases, offering a valuable tool for HPLC method development. These columns feature an embedded carbamate group in the bonded phase ligand, which provides alternate selectivity, particularly for phenolic compounds, when compared to traditional straight-chain alkyl columns.
  • Alternate selectivity, especially for phenolic compounds
  • Embedded carbamate group in the bonded phase ligand
  • Complements C18 and C8 phases
  • Embedded polar C18 providing different selectivity with improved peak shape for basic compounds
Catalog No. 50-782-070 Supplier Waters Corp Supplier No. 186003935
Add to Cart
Add to Cart
Request a Quote
The XBridge BEH Shield RP18 columns are designed to complement XBridge BEH C18 and C8 phases, offering a valuable tool for HPLC method development. These columns feature an embedded carbamate group in the bonded phase ligand, which provides alternate selectivity, particularly for phenolic compounds, when compared to traditional straight-chain alkyl columns.
  • Alternate selectivity, especially for phenolic compounds
  • Embedded carbamate group in the bonded phase ligand
  • Complements C18 and C8 phases
  • Embedded polar C18 providing different selectivity with improved peak shape for basic compounds