Learn More
Waters Corp XBridge OBD Prep Column, 130Å, 10 μm, 30 mm X 150 mm
Catalog No. 50782118 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
BEH Shield RP18 columns complement BEH C18 and C8 phases for HPLC method development. The embedded carbamate group provides alternate selectivity for phenolic compounds.
  • Complements BEH C18 and C8 phases for HPLC method development.
  • Embedded carbamate group offers alternate selectivity for phenolic compounds.
  • Designed for preparative HPLC and method development.
  • Features innovative Optimum Bed Density (OBD) column format for direct scalability.
  • MaxPeak High-Performance Surface Technology prevents unwanted analyte-surface interactions.
  • Wide, usable pH range accommodates varying analytes.
  • Maintains superior chemical stability across varying analytes.
  • Utilizes BEH base particle for mechanical and chemical stability.
  • Available in multiple column chemistries (C18, C8, HILIC, Amide).
  • Hazardous material: special freight handling and charges may apply.
Catalog No. 50-782-118 Supplier Waters Corp Supplier No. 186003996
Add to Cart
Add to Cart
Request a Quote
BEH Shield RP18 columns complement BEH C18 and C8 phases for HPLC method development. The embedded carbamate group provides alternate selectivity for phenolic compounds.
  • Complements BEH C18 and C8 phases for HPLC method development.
  • Embedded carbamate group offers alternate selectivity for phenolic compounds.
  • Designed for preparative HPLC and method development.
  • Features innovative Optimum Bed Density (OBD) column format for direct scalability.
  • MaxPeak High-Performance Surface Technology prevents unwanted analyte-surface interactions.
  • Wide, usable pH range accommodates varying analytes.
  • Maintains superior chemical stability across varying analytes.
  • Utilizes BEH base particle for mechanical and chemical stability.
  • Available in multiple column chemistries (C18, C8, HILIC, Amide).
  • Hazardous material: special freight handling and charges may apply.