Learn More
Waters Corp Waters XBridge Protein BEH C4 Column, 300Å, 2.5 μm
Catalog No. 502038915 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • The XBridge Protein BEH C4 Column is optimized and quality control (QC) tested for protein separations using a MassPREP Protein Mix standard. Its 300Å pore size trifunctionally bonded BEH particle offers a wide usable pH range, superior low pH stability, and ultra-low column bleed, making it suitable for various protein-based applications. It is suggested for low-level intact protein and subunit analysis.
  • Optimized and QC tested for protein separations.
  • 300Å pore size.
  • Trifunctionally bonded BEH particle.
  • Offers the widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed for protein based applications.
  • Suitable for low-level intact protein and subunit analysis.
Catalog No. 50-203-8915 Supplier Waters Corp Supplier No. 186009129
  • The XBridge Protein BEH C4 Column is optimized and quality control (QC) tested for protein separations using a MassPREP Protein Mix standard. Its 300Å pore size trifunctionally bonded BEH particle offers a wide usable pH range, superior low pH stability, and ultra-low column bleed, making it suitable for various protein-based applications. It is suggested for low-level intact protein and subunit analysis.
  • Optimized and QC tested for protein separations.
  • 300Å pore size.
  • Trifunctionally bonded BEH particle.
  • Offers the widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed for protein based applications.
  • Suitable for low-level intact protein and subunit analysis.