Learn More
Waters Corp Waters XBridge Protein BEH C4 Guard Column, 300Å, 5 μm, 2.1 mm X 30 mm
Catalog No. 502038963 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Protein BEH C4 Guard Column supports reversed-phase protein separation. It features a C4 ligand to reduce retention and improve peak capacity.
  • Designed for protein separation.
  • Utilizes BEH particle technology.
  • Features a C4 ligand for reduced retention.
  • Minimizes protein carryover and increases protein recovery.
  • Optimized for improved peak capacity.
  • QC tested for consistent performance.
  • Compatible with UPLC systems.
  • Operates effectively across a wide pH range of 1 to 12.
  • Withstands temperatures up to 90°C.
  • Maximum pressure tolerance of 6000 psi (415 Bar).
  • Low silanol activity ensures reliable results.
  • Suitable for molecular weights ranging from 1000 to 500000.
  • Provided as a single unit per package.
Catalog No. 50-203-8963 Supplier Waters Corp Supplier No. 186009269
The XBridge Protein BEH C4 Guard Column supports reversed-phase protein separation. It features a C4 ligand to reduce retention and improve peak capacity.
  • Designed for protein separation.
  • Utilizes BEH particle technology.
  • Features a C4 ligand for reduced retention.
  • Minimizes protein carryover and increases protein recovery.
  • Optimized for improved peak capacity.
  • QC tested for consistent performance.
  • Compatible with UPLC systems.
  • Operates effectively across a wide pH range of 1 to 12.
  • Withstands temperatures up to 90°C.
  • Maximum pressure tolerance of 6000 psi (415 Bar).
  • Low silanol activity ensures reliable results.
  • Suitable for molecular weights ranging from 1000 to 500000.
  • Provided as a single unit per package.