Learn More
Waters Corp XBridge Protein BEH C4 OBD Prep Column, 300 Å, 5 μm, 19 x 150 mm
Catalog No. 502390839 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is QC tested and optimized to separate proteins based on size, hydrophobicity, and isoelectric point. Compared to traditional C18 phases, its C4 ligand is less retentive, which minimizes protein carryover, increases protein recovery, and improves peak capacity.
  • Optimized for protein separation based on size, hydrophobicity, and isoelectric point.
  • Less retentive C4 ligand minimizes protein carryover.
  • Increases protein recovery.
  • Improves peak capacity.
  • Features a trifunctional C4 ligand, fully end-capped.
  • Utilizes Ethylene-Bridged Hybrid (BEH) particles.
  • QC tested with complex protein standard mixtures for reproducible performance.
Catalog No. 50-239-0839 Supplier Waters Corp Supplier No. 186007313
Add to Cart
Add to Cart
This column is QC tested and optimized to separate proteins based on size, hydrophobicity, and isoelectric point. Compared to traditional C18 phases, its C4 ligand is less retentive, which minimizes protein carryover, increases protein recovery, and improves peak capacity.
  • Optimized for protein separation based on size, hydrophobicity, and isoelectric point.
  • Less retentive C4 ligand minimizes protein carryover.
  • Increases protein recovery.
  • Improves peak capacity.
  • Features a trifunctional C4 ligand, fully end-capped.
  • Utilizes Ethylene-Bridged Hybrid (BEH) particles.
  • QC tested with complex protein standard mixtures for reproducible performance.