Learn More
Waters Corp XBridge Protein BEH C4 OBD Prep Column, 300 Å, 10 μm, 10 mm × 50 mm
Catalog No. 502391629 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XBridge Protein BEH C4 OBD Prep Column is designed to separate proteins based on size, hydrophobicity, and isoelectric point. Its C4 ligand offers less retention compared to traditional C18 phases, which helps minimize protein carryover, increase protein recovery, and improve peak capacity.
  • Optimized for protein separation.
  • Trifunctional C4 ligand on BEH particles.
  • Less retentive C4 minimizes protein carryover, increases recovery, and improves peak capacity.
  • Batch quality control tested with protein standards.
  • Uniform packing for optimum density.
  • Direct scalability from analytical to preparative.
  • Excellent column lifetime and consistent performance.
  • Good resolution, peak shape, and high mass loading.
  • Maximizes recoveries for targeted small molecules and biomolecules.
Catalog No. 50-239-1629 Supplier Waters Corp Supplier No. 186008276
This XBridge Protein BEH C4 OBD Prep Column is designed to separate proteins based on size, hydrophobicity, and isoelectric point. Its C4 ligand offers less retention compared to traditional C18 phases, which helps minimize protein carryover, increase protein recovery, and improve peak capacity.
  • Optimized for protein separation.
  • Trifunctional C4 ligand on BEH particles.
  • Less retentive C4 minimizes protein carryover, increases recovery, and improves peak capacity.
  • Batch quality control tested with protein standards.
  • Uniform packing for optimum density.
  • Direct scalability from analytical to preparative.
  • Excellent column lifetime and consistent performance.
  • Good resolution, peak shape, and high mass loading.
  • Maximizes recoveries for targeted small molecules and biomolecules.