Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XBridge Protein BEH C4 OBD Column, 300 Å, 10 μm, 10 × 150 mm

Supplier:  Waters Corp 186008278

Encompass_Preferred

This XBridge Protein BEH C4 OBD Column is designed for preparative chromatography applications involving proteins.

  • Dimension: 10 × 150 mm
  • Particle size: 10 μm
  • Chemistry: XBridge Protein BEH C4, 300 Å

Catalog No. 50-239-1981


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.