Learn More
Waters Corp XBridge Protein BEH C4 OBD Prep Column, 300 Å, 10 μm, 19 mm x 100 mm
Catalog No. 502390848 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This OBD Prep Column is designed for high-performance protein separation. It features a trifunctional C4 ligand bonded to Ethylene-Bridged Hybrid (BEH) particles, offering less retention than traditional C18 phases.
  • QC tested and optimized to separate proteins based on size, hydrophobicity, and isoelectric point.
  • Less retentive compared to traditional C18 phases.
  • Minimizes protein carryover.
  • Increases protein recovery.
  • Improves peak capacity.
  • Trifunctional C4 ligand, fully end-capped, and bonded to Ethylene-Bridged Hybrid (BEH) particles.
  • Each synthesized batch is specifically QC tested with a complex mixture of protein standards to help ensure reproducible Prep Column Performance.
Catalog No. 50-239-0848 Supplier Waters Corp Supplier No. 186007332
This OBD Prep Column is designed for high-performance protein separation. It features a trifunctional C4 ligand bonded to Ethylene-Bridged Hybrid (BEH) particles, offering less retention than traditional C18 phases.
  • QC tested and optimized to separate proteins based on size, hydrophobicity, and isoelectric point.
  • Less retentive compared to traditional C18 phases.
  • Minimizes protein carryover.
  • Increases protein recovery.
  • Improves peak capacity.
  • Trifunctional C4 ligand, fully end-capped, and bonded to Ethylene-Bridged Hybrid (BEH) particles.
  • Each synthesized batch is specifically QC tested with a complex mixture of protein standards to help ensure reproducible Prep Column Performance.