Learn More
Waters Corp HPLC Guard Cartridges, 300Å, 10 μm, 10 x 10 mm
Supplier:  Waters Corp 186007325
Encompass_Preferred
The XBridge Protein BEH C4 Prep Guard Cartridge is an HPLC guard cartridge with a 300Å pore size, 10 μm particle size, and dimensions of 10 mm length by 10 mm internal diameter. It features C4 chemistry with BEH technology, designed for reversed-phase chromatography.
  • Hplc guard cartridges
  • 10 mm length
  • 10 mm internal diameter
  • 10 μm particle size
  • C4 chemistry
  • Beh C4 chemistry producers term
  • 300 Å pore size
  • Usp class L26
  • 8.0 % carbon load
  • 90.0 m2/g bet surface area
  • Non endcapped
  • Made in the United States of America
  • Stainless steel hardware material
  • Ph range of 2.0 to 10.0
  • Reversed phase chromatography separation mode
  • Hybrid matrix
  • Spherical, fully porous particle morphology
Catalog No. 50-239-1734