Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Prep Guard Cartridge, 300 Å, 10 μm, 30 mm x 10 mm

Catalog No. 502392183 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

XBridge Protein BEH C4 Prep Guard Cartridges are QC tested and optimized for separating proteins by size, hydrophobicity, and isoelectric point. The C4 ligand offers less retention than traditional C18 phases, minimizing protein carryover and enhancing recovery and peak capacity.

  • Chemistry: C4
  • Separation mode: Reversed phase
  • Particle substrate: Hybrid
  • pH range: 1 to 10
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped: No
  • Silanol activity: Low
  • Maximum molecular weight range: 150000
  • Particle shape: Spherical
  • Particle size: 10 μm
  • Pore size: 300 Å
  • QC tested for protein
  • Surface area: 90
  • System: HPLC
  • Particle technology: BEH
  • USP classification: L26
  • Inner diameter: 30 mm
  • Length: 10 mm
  • Carbon load: 8 %
  • Application: Protein

Catalog No. 50-239-2183 Supplier Waters Corp Supplier No. 186007335
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XBridge Protein BEH C4 Prep Guard Cartridges are QC tested and optimized for separating proteins by size, hydrophobicity, and isoelectric point. The C4 ligand offers less retention than traditional C18 phases, minimizing protein carryover and enhancing recovery and peak capacity.

  • Chemistry: C4
  • Separation mode: Reversed phase
  • Particle substrate: Hybrid
  • pH range: 1 to 10
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped: No
  • Silanol activity: Low
  • Maximum molecular weight range: 150000
  • Particle shape: Spherical
  • Particle size: 10 μm
  • Pore size: 300 Å
  • QC tested for protein
  • Surface area: 90
  • System: HPLC
  • Particle technology: BEH
  • USP classification: L26
  • Inner diameter: 30 mm
  • Length: 10 mm
  • Carbon load: 8 %
  • Application: Protein

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.