Learn More
Waters Corp XBridge Prep Guard Cartridge, 300 Å, 5 μm, 19 x 10 mm
Supplier:  Waters Corp 186007310
Encompass_Preferred
The XBridge Protein BEH C4 Prep Guard Cartridge is optimized for protein separation, leveraging a C4 ligand that is less retentive than traditional C18 phases. This design minimizes protein carryover, enhances protein recovery, and improves peak capacity, making it ideal for protecting columns used in critical applications.
  • QC tested and optimized for protein separation based on size, hydrophobicity, and isoelectric point.
  • C4 ligand minimizes protein carryover compared to traditional C18 phases.
  • Increases protein recovery during separation.
  • Improves peak capacity for better analytical results.
Catalog No. 50-239-0499