Learn More
Waters Corp Xbridge Protein BEH C4 VanGuard Cartridge, 300Å, 2.5 µm, 2.1 mm X 5 mm, 3/pk
Catalog No. NC1868762 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Protein BEH C4 300Å columns are optimized and QC tested for protein separations. The 300Å pore size trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for various protein based applications. This Part Number is for the appropriate 2.1 x 5 mm VanGaurd Cartridge, designed to protect the 2.1 mm I.D. analytical column, and requires the reuable Universal Guard Cartridge Holder (Part Number 186007949) for use.
Catalog No. NC1868762 Supplier Waters Corp Supplier No. 186009131
The XBridge Protein BEH C4 300Å columns are optimized and QC tested for protein separations. The 300Å pore size trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for various protein based applications. This Part Number is for the appropriate 2.1 x 5 mm VanGaurd Cartridge, designed to protect the 2.1 mm I.D. analytical column, and requires the reuable Universal Guard Cartridge Holder (Part Number 186007949) for use.