Learn More
Waters Corp Column, BEH, 2.5 μm, 125 Å, 4.6 mm x 150 mm
Catalog No. 502391910 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column contains 2.5μm particles based on Ethylene Bridged Hybrid (BEH) particle technology and a diol-bonded surface coating, similar to those used in HPLC-based SEC columns. It is designed for separating proteins ranging from 1,000 to 80,000 Daltons.
  • Based on Ethylene Bridged Hybrid (BEH) particle technology.
  • Utilizes diol-bonded surface coating.
  • Suitable for separating proteins ranging from 1,000 to 80,000 Daltons.
  • Includes a SEC protein standard.
Catalog No. 50-239-1910 Supplier Waters Corp Supplier No. 176004332
This column contains 2.5μm particles based on Ethylene Bridged Hybrid (BEH) particle technology and a diol-bonded surface coating, similar to those used in HPLC-based SEC columns. It is designed for separating proteins ranging from 1,000 to 80,000 Daltons.
  • Based on Ethylene Bridged Hybrid (BEH) particle technology.
  • Utilizes diol-bonded surface coating.
  • Suitable for separating proteins ranging from 1,000 to 80,000 Daltons.
  • Includes a SEC protein standard.