Learn More
Waters Corp XBridge Protein BEH SEC Guard Column, 450 Å, 2.5 μm, 4.6 mm x 150 mm
Supplier:  Waters Corp 176004338
Encompass_Preferred
This XBridge Protein BEH SEC 450Å Guard Column utilizes Ethylene Bridged Hybrid (BEH)-based particle technology and a diol-bonded surface coating. It is designed for separating proteins ranging from 100K to 1,500K Daltons.
  • Utilizes Ethylene Bridged Hybrid (BEH)-based particle technology.
  • Diol-bonded surface coating.
  • Suitable for separating proteins from 100K to 1,500K Daltons.
  • Includes one vial of the BEH450 Protein Std.
Catalog No. 50-239-2070