Learn More
Waters Corp XBridge Protein BEH SEC Column, 450Å, 2.5 μm, 4.6 x 300 mm
Supplier:  Waters Corp 186009180
Encompass_Preferred
The XBridge Protein BEH SEC Columns complement UPLC-based SEC offerings, providing an option for traditional HPLC-based instrumentation in peptide or protein size-exclusion chromatography (SEC). These columns use Ethylene Bridged Hybrid (BEH) particle technology and a diol-bonded surface coating, enabling method transfer based on lab instrumentation, component resolution, or sample throughput needs.
  • Utilizes Ethylene Bridged Hybrid (BEH) particle technology.
  • Features a diol-bonded surface coating.
  • Manufactured in a cGMP, ISO 9001 certified plant with stringent protocols and ultra-pure reagents.
  • Each batch undergoes standard QC measurements and an application-specific test to ensure reproducible performance.
  • Enables easy method transferability between different lab instrumentation.
  • Maximum suggested back pressure: 5000 PSI (345 Bar).
Catalog No. 50-239-0464