Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Guard Column, SEC, 450 A, 3.5 um, 7.8 mm ID

Catalog No. 502390561 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This guard column is built upon ethylene bridged hybrid (BEH)-based particle technology and diol-bonded surface coating. It is optimally suited for separating proteins with molecular weights ranging from 100,000 to 1,500,000 Daltons.

  • Designed for high-speed applications in UPLC systems.
  • Features SEC chemistry and gel filtration (aqueous) separation mode.
  • Particle substrate is hybrid with spherical shape.
  • Optimized for a pH range from 2.5 to 8.
  • Maximum operating temperature is 60 °C.
  • Withstands a maximum pressure of 3000 psi (207 bar).
  • Low silanol activity.
  • Pore size of 450 Å and particle size of 3.5 μm.
  • Packed with 20% methanol in water solvent.
  • Surface area of 80.
  • Inner diameter of 7.8 mm and length of 30 mm.
  • Carbon load of 9%.
  • Ships with one vial of BEC450 SEC protein standard at no additional cost.

Catalog No. 50-239-0561 Supplier Waters Corp Supplier No. 176003597
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This guard column is built upon ethylene bridged hybrid (BEH)-based particle technology and diol-bonded surface coating. It is optimally suited for separating proteins with molecular weights ranging from 100,000 to 1,500,000 Daltons.

  • Designed for high-speed applications in UPLC systems.
  • Features SEC chemistry and gel filtration (aqueous) separation mode.
  • Particle substrate is hybrid with spherical shape.
  • Optimized for a pH range from 2.5 to 8.
  • Maximum operating temperature is 60 °C.
  • Withstands a maximum pressure of 3000 psi (207 bar).
  • Low silanol activity.
  • Pore size of 450 Å and particle size of 3.5 μm.
  • Packed with 20% methanol in water solvent.
  • Surface area of 80.
  • Inner diameter of 7.8 mm and length of 30 mm.
  • Carbon load of 9%.
  • Ships with one vial of BEC450 SEC protein standard at no additional cost.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.