Learn More
Waters Corp SEC Column, 200Å, 2.5 μm, 4.6 mm x 300 mm, BEH
Supplier:  Waters Corp 176004336
Encompass_Preferred
These SEC Columns, with 200Å and 2.5μm Ethylene Bridged Hybrid (BEH) particles and diol-bonded surface, separate proteins from 10,000 to 450,000 Daltons. Includes a BEH200 SEC Protein Standard Mix vial. Manufactured to ISO9001/cGMP, they ensure mobile-phase pH stability and assay reliability.
  • BEH-based particle technology with diol-bonded surface.
  • Separates proteins from 10,000 to 450,000 Daltons.
  • Includes BEH200 SEC Protein Standard Mix vial.
  • Exceptional mobile-phase pH stability for extended column lifespan.
  • Suitable for QC or advanced LC-MS assays.
  • Manufactured to ISO9001/cGMP.
  • Delivers robust assays, unparalleled peak particle shape, capacity, efficiency, and reliability.
  • Optimal conditions: pH 2.5 to 8, max temp 60°C, max pressure 6,500 psi (448 Bar).
Catalog No. 50-239-1942