Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XBridge Protein BEH SEC Column, 125Å, 2.5μm, 7.8 mm × 150 mm

Catalog No. 502392318 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XBridge Protein BEH SEC Column, 125 Å, contains 2.5μm particles and is based on Ethylene Bridged Hybrid (BEH)-based particle technology with a diol-bonded surface coating. These columns are designed for HPLC-based size exclusion chromatography and are well-suited to separate proteins ranging from 1,000 to 80,000 Daltons.

  • Separation mode: Gel filtration (aqueous)
  • Particle substrate: Hybrid
  • pH range: 2.5 to 8 pH
  • Temperature limit: 60°C
  • Maximum pressure: 5500 psi (379 Bar)
  • Pore size: 125Å
  • Particle size: 2.5μm
  • Inner diameter: 7.8 mm
  • Length: 150 mm
  • Bonding technology: Diol (OH)
  • Silanol activity: Low
  • Molecular weight range: 1,000 to 80,000
  • Endfitting type: Parker-style
  • Packing solvent: 20% methanol in water
  • Application: Protein

Catalog No. 50-239-2318 Supplier Waters Corp Supplier No. 176004323
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XBridge Protein BEH SEC Column, 125 Å, contains 2.5μm particles and is based on Ethylene Bridged Hybrid (BEH)-based particle technology with a diol-bonded surface coating. These columns are designed for HPLC-based size exclusion chromatography and are well-suited to separate proteins ranging from 1,000 to 80,000 Daltons.

  • Separation mode: Gel filtration (aqueous)
  • Particle substrate: Hybrid
  • pH range: 2.5 to 8 pH
  • Temperature limit: 60°C
  • Maximum pressure: 5500 psi (379 Bar)
  • Pore size: 125Å
  • Particle size: 2.5μm
  • Inner diameter: 7.8 mm
  • Length: 150 mm
  • Bonding technology: Diol (OH)
  • Silanol activity: Low
  • Molecular weight range: 1,000 to 80,000
  • Endfitting type: Parker-style
  • Packing solvent: 20% methanol in water
  • Application: Protein

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.