Learn More
Waters Corp XBridge Protein BEH SEC Column, 125Å, 2.5 μm, 4.6 mm ID
Catalog No. NC3869746 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Protein BEH SEC Column, 125Å, 2.5 μm, 4.6 × 300 mm, contains 2.5 μm particles based on Ethylene Bridged Hybrid (BEH)-based particle technology and diol-bonded surface coating. These columns are designed for the separation of proteins ranging from 1,000 to 80,000 Daltons. Each batch undergoes specific QC testing to ensure consistent performance for challenging size-based protein separations.
  • Based on Ethylene Bridged Hybrid (BEH)-based particle technology.
  • Utilizes diol-bonded surface coating.
  • Suitable for separating proteins in the molecular weight range of 1,000 to 80,000 Daltons.
  • Each batch is QC tested for consistent performance.
  • Available with a vial of BEH125 Protein Standard at no additional charge.
  • Suggested uses include Gel Filtration, Size Exclusion Chromatography, Aggregate, Monomer, and Fragment analysis.
Catalog No. NC3869746 Supplier Waters Corp Supplier No. 186009172
The XBridge Protein BEH SEC Column, 125Å, 2.5 μm, 4.6 × 300 mm, contains 2.5 μm particles based on Ethylene Bridged Hybrid (BEH)-based particle technology and diol-bonded surface coating. These columns are designed for the separation of proteins ranging from 1,000 to 80,000 Daltons. Each batch undergoes specific QC testing to ensure consistent performance for challenging size-based protein separations.
  • Based on Ethylene Bridged Hybrid (BEH)-based particle technology.
  • Utilizes diol-bonded surface coating.
  • Suitable for separating proteins in the molecular weight range of 1,000 to 80,000 Daltons.
  • Each batch is QC tested for consistent performance.
  • Available with a vial of BEH125 Protein Standard at no additional charge.
  • Suggested uses include Gel Filtration, Size Exclusion Chromatography, Aggregate, Monomer, and Fragment analysis.