Learn More
Waters Corp WATERS CORPORATION XBridge Protein BEH SEC Column, 200Å, 3.5 μm, 7.8 mm x 150 mm
Catalog No. 502038133 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge Protein BEH SEC, 200Å Columns, containing 3.5μm particles are based on the same Waters Ethylene Bridged Hybrid (BEH)-based particle technology and diol-bonded surface coating as used in Waters' successful line of HPLC-based SEC columns. They are best suited to separate proteins ranging from 10,000–450,000 Daltons.
  • Based on Ethylene Bridged Hybrid (BEH)-based particle technology and diol-bonded surface coating.
  • Separates proteins ranging from 10,000 to 450,000 Daltons.
Catalog No. 50-203-8133 Supplier Waters Corp Supplier No. 186007639
XBridge Protein BEH SEC, 200Å Columns, containing 3.5μm particles are based on the same Waters Ethylene Bridged Hybrid (BEH)-based particle technology and diol-bonded surface coating as used in Waters' successful line of HPLC-based SEC columns. They are best suited to separate proteins ranging from 10,000–450,000 Daltons.
  • Based on Ethylene Bridged Hybrid (BEH)-based particle technology and diol-bonded surface coating.
  • Separates proteins ranging from 10,000 to 450,000 Daltons.