Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters XBridge Protein BEH SEC Column, 200Å, 2.5 μm

Catalog No. 502038925 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XBridge Protein BEH SEC Column, 200Å, 2.5 μm, 7.8 mm x 150 mm, contains 2.5 μm particles with Ethylene Bridged Hybrid (BEH)-based technology and diol-bonded surface coating. These columns are designed to separate proteins ranging from 10,000 to 450,000 Daltons, with each batch QC tested for consistent performance in size-based protein separations.

  • Increases productivity for LC-MS assays and quality-control procedures.
  • Offers unique mobile-phase pH stability, extended column lifetime, and great column dependability.
  • Provides exceptional particle efficiency and peak form.
  • Allows seamless migration to UPLC technology.
  • Manufactured adhering to ISO9001 and cGMP guidelines, with consistent QC testing.
  • Optimal pH range: 2 to 8.
  • Temperature limits: 60 °C.
  • Maximum pressure: 4500 psi (310 bar).
  • System compatibility: UHPLC, UPLC, HPLC.

Catalog No. 50-203-8925 Supplier Waters Corp Supplier No. 186009163
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XBridge Protein BEH SEC Column, 200Å, 2.5 μm, 7.8 mm x 150 mm, contains 2.5 μm particles with Ethylene Bridged Hybrid (BEH)-based technology and diol-bonded surface coating. These columns are designed to separate proteins ranging from 10,000 to 450,000 Daltons, with each batch QC tested for consistent performance in size-based protein separations.

  • Increases productivity for LC-MS assays and quality-control procedures.
  • Offers unique mobile-phase pH stability, extended column lifetime, and great column dependability.
  • Provides exceptional particle efficiency and peak form.
  • Allows seamless migration to UPLC technology.
  • Manufactured adhering to ISO9001 and cGMP guidelines, with consistent QC testing.
  • Optimal pH range: 2 to 8.
  • Temperature limits: 60 °C.
  • Maximum pressure: 4500 psi (310 bar).
  • System compatibility: UHPLC, UPLC, HPLC.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.