Learn More
Waters Corp Waters Corporation XBridge Protein BEH SEC Column, 200Å, 2.5 μm, 7.8 mm X 300 mm
Supplier:  Waters Corp 186009164
Encompass_Preferred
The XBridge Protein BEH SEC Column, 200Å, 2.5 μm, 7.8 mm X 300 mm, features Ethylene Bridged Hybrid (BEH) particle technology and a diol-bonded surface. It separates proteins from 10,000 to 450,000 Daltons and is QC tested with BEH200 SEC Protein Standard for consistent performance in gel filtration, size exclusion chromatography, and aggregate/monomer/fragment analysis.
  • Longer column life compared to silica-based SEC columns.
  • Enhanced sample throughput and HPLC-based SEC resolution due to flow and pressure tolerance.
  • Manufactured in cGMP, ISO9001, and ISO13485 certified facilities.
  • Performs within an operating pH range of 2.5-8.
Catalog No. 50-203-8926