Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XBridge Protein BEH SEC Column, 200Å, 2.5 μm, 4.6 mm X 300 mm

Catalog No. NC3806173 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This XBridge Protein BEH SEC Column features 2.5 μm Ethylene Bridged Hybrid (BEH) particles and a diol-bonded surface coating. It is designed for separating proteins from 10,000 to 450,000 Daltons, offering consistent performance, exceptional pH stability, and great particle efficiency. This column is suitable for LC-MS assays, quality-control, and UPLC technology transitions.

  • Particle size: 2.5 μm
  • Pore size: 200 Å
  • Dimensions: 4.6 mm X 300 mm
  • Molecular weight range: 10,000 to 450,000 Daltons
  • pH range: 2.5 to 8
  • Temperature limits: 60°C
  • Maximum pressure: 6,500 psi (448 Bar)
  • Compatible with UHPLC, UPLC, and HPLC systems

Catalog No. NC3806173 Supplier Waters Corp Supplier No. 186009176
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This XBridge Protein BEH SEC Column features 2.5 μm Ethylene Bridged Hybrid (BEH) particles and a diol-bonded surface coating. It is designed for separating proteins from 10,000 to 450,000 Daltons, offering consistent performance, exceptional pH stability, and great particle efficiency. This column is suitable for LC-MS assays, quality-control, and UPLC technology transitions.

  • Particle size: 2.5 μm
  • Pore size: 200 Å
  • Dimensions: 4.6 mm X 300 mm
  • Molecular weight range: 10,000 to 450,000 Daltons
  • pH range: 2.5 to 8
  • Temperature limits: 60°C
  • Maximum pressure: 6,500 psi (448 Bar)
  • Compatible with UHPLC, UPLC, and HPLC systems

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.