Learn More
Waters Corp Waters Corporation XBridge Protein BEH SEC Column, 450 Å, 3.5 µm, 7.8 mm X 150 mm
Catalog No. 502037425 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Protein BEH SEC Column is designed for peptide or protein size-exclusion chromatography (SEC) using HPLC. It uses Ethylene Bridged Hybrid (BEH) particle technology and a diol-bonded surface coating, optimizing it for separating proteins from 100,000 to 1,500,000 Daltons.
  • BEH particle technology with diol-bonded surface.
  • Separates proteins from 100,000 to 1,500,000 Daltons.
  • Includes a BEH450 SEC Protein Standard Mix.
  • Improved flow and pressure tolerance for high throughput.
  • Supports UPLC/HPLC/UHPLC-based SEC method transfer.
  • Manufactured in cGMP, ISO9001, and ISO13485 facilities.
  • Exceptional longevity across wide pH and temperature ranges.
  • Operates in 2.5–8 pH, up to 60 °C, and max 3000 PSI (207 Bar).
  • Ensures reproducible batch-to-batch performance.
Catalog No. 50-203-7425 Supplier Waters Corp Supplier No. 176003598
The XBridge Protein BEH SEC Column is designed for peptide or protein size-exclusion chromatography (SEC) using HPLC. It uses Ethylene Bridged Hybrid (BEH) particle technology and a diol-bonded surface coating, optimizing it for separating proteins from 100,000 to 1,500,000 Daltons.
  • BEH particle technology with diol-bonded surface.
  • Separates proteins from 100,000 to 1,500,000 Daltons.
  • Includes a BEH450 SEC Protein Standard Mix.
  • Improved flow and pressure tolerance for high throughput.
  • Supports UPLC/HPLC/UHPLC-based SEC method transfer.
  • Manufactured in cGMP, ISO9001, and ISO13485 facilities.
  • Exceptional longevity across wide pH and temperature ranges.
  • Operates in 2.5–8 pH, up to 60 °C, and max 3000 PSI (207 Bar).
  • Ensures reproducible batch-to-batch performance.