Learn More
Waters Corp Waters XBridge Protein BEH SEC Column, 125 Å, 3.5 μm, 7.8 mm × 300 mm
Catalog No. NC1269452 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge Protein BEH SEC, 125 Å Columns feature 3.5μm particles, built on Ethylene Bridged Hybrid (BEH)-based particle technology and diol-bonded surface coating. They are designed for separating proteins from 1,000 to 80,000 Daltons, enabling streamlined method development and transfer for HPLC-based SEC applications.
  • Complements existing smaller particle size UPLC-based SEC columns.
  • Exceptional column life with reduced undesirable protein and column interactions.
  • Provides HPLC-based SEC resolution of proteins from 10,000 to 1,500,000 Daltons with high throughput.
  • Offers flow and pressure tolerance for increased sample throughput on HPLC systems.
  • BEH base particle technology and innovative diol bonding ensure column stability, performance, and lifetime.
Catalog No. NC1269452 Supplier Waters Corp Supplier No. 186007637
XBridge Protein BEH SEC, 125 Å Columns feature 3.5μm particles, built on Ethylene Bridged Hybrid (BEH)-based particle technology and diol-bonded surface coating. They are designed for separating proteins from 1,000 to 80,000 Daltons, enabling streamlined method development and transfer for HPLC-based SEC applications.
  • Complements existing smaller particle size UPLC-based SEC columns.
  • Exceptional column life with reduced undesirable protein and column interactions.
  • Provides HPLC-based SEC resolution of proteins from 10,000 to 1,500,000 Daltons with high throughput.
  • Offers flow and pressure tolerance for increased sample throughput on HPLC systems.
  • BEH base particle technology and innovative diol bonding ensure column stability, performance, and lifetime.