Learn More
Waters Corp Waters XBridge Protein BEH SEC Column, 125Å, 2.5μm, 7.8 mm × 300 mm
Supplier: Waters Corp 176004324
This SEC column contains 2.5 μm particles and is based on Ethylene Bridged Hybrid (BEH)-based particle technology with a diol-bonded surface coating. It is designed for separating proteins with molecular weights ranging from 1,000 to 80,000 Daltons, offering compatibility with HPLC systems. This product also includes one vial of the BEH125 SEC Protein Std.
- Chemistry: SEC (Size Exclusion Chromatography).
- Separation mode: Gel filtration (aqueous).
- Particle substrate: Hybrid.
- pH range: 2.5 to 8.
- Temperature limits: 60 °C.
- Maximum pressure: 7500 psi (517 Bar).
- Pore size: 125 Å.
- Inner diameter: 7.8 mm.
- Length: 300 mm.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.