Learn More
Waters Corp Waters XBridge Protein BEH SEC Column, 200Å, 2.5 μm, 7.8 mm X 300 mm
Catalog No. 502037582 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Protein BEH SEC, 200 Å Column, with 2.5 μm particles, uses Ethylene Bridged Hybrid (BEH)-based particle technology and diol-bonded surface coating. It is ideal for separating proteins from 10,000 to 450,000 Daltons.
  • Based on Ethylene Bridged Hybrid (BEH)-based particle technology and diol-bonded surface coating.
  • Separates proteins from 10,000 to 450,000 Daltons.
  • Includes one vial of BEH200 SEC Protein Standard Mix.
  • Facilitates easy migration to UltraPerformance LC technology.
  • Offers unrivaled column longevity and versatility in mobile phase pH and temperature ranges.
  • Improves pH stability for longer column lives.
  • Enhances particle efficiency for good peak shape and peak capacity.
  • Leverages assay robustness and improves column reliability.
Catalog No. 50-203-7582 Supplier Waters Corp Supplier No. 176004327
The XBridge Protein BEH SEC, 200 Å Column, with 2.5 μm particles, uses Ethylene Bridged Hybrid (BEH)-based particle technology and diol-bonded surface coating. It is ideal for separating proteins from 10,000 to 450,000 Daltons.
  • Based on Ethylene Bridged Hybrid (BEH)-based particle technology and diol-bonded surface coating.
  • Separates proteins from 10,000 to 450,000 Daltons.
  • Includes one vial of BEH200 SEC Protein Standard Mix.
  • Facilitates easy migration to UltraPerformance LC technology.
  • Offers unrivaled column longevity and versatility in mobile phase pH and temperature ranges.
  • Improves pH stability for longer column lives.
  • Enhances particle efficiency for good peak shape and peak capacity.
  • Leverages assay robustness and improves column reliability.