Learn More
Waters Corp XBridge Protein BEH SEC Guard Column, 450Å, 2.5 μm, 4.6 mm X 30 mm
Supplier:  Waters Corp 176004337
Encompass_Preferred
The Waters XBridge Protein BEH SEC Guard Column, 450 Å, with 2.5 μm particles, is engineered to separate proteins ranging from 100K to 1,500K Daltons. It utilizes Ethylene Bridged Hybrid (BEH)-based particle technology and diol-bonded surface coating to ensure consistent performance and extend analytical column lifetime. Each package includes one vial of BEH450 Protein Std (P/N 186006519).
  • Enhanced mobile-phase pH stability (2 to 8 pH).
  • Separates proteins from 100K to 1,500K Daltons.
  • Features BEH-based particle technology.
  • Diol-bonded surface coating.
  • Manufactured under ISO9001 and cGMP guidelines.
  • Includes BEH450 Protein Std.
Catalog No. 50-239-0964