Learn More
Waters Corp XBridge Protein BEH SEC Guard Column, 200Å, 2.5 μm
Catalog No. 502037580 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Protein BEH SEC, 200 Å Guard Column, with 2.5 μm particles, utilizes Waters Ethylene Bridged Hybrid (BEH) technology and diol-bonded surface coating. Ideal for separating proteins from 10,000 to 450,000 Daltons, it offers mobile-phase pH stability for extended column lifetime. Suitable for quality control or LC-MS assays, it ensures durability, assay robustness, and exceptional efficiency.
  • Utilizes ethylene bridged hybrid (BEH)-based particle technology with diol-bonded surface coating.
  • Separates proteins within a 10,000 to 450,000 Dalton molecular weight range.
  • Ensures mobile-phase pH stability, extending column lifetime.
  • Supports quality control procedures and LC-MS assays.
  • Delivers column durability, assay robustness, and exceptional efficiency.
  • Includes one vial of protein standard at no additional cost.
Catalog No. 50-203-7580 Supplier Waters Corp Supplier No. 176004325
The XBridge Protein BEH SEC, 200 Å Guard Column, with 2.5 μm particles, utilizes Waters Ethylene Bridged Hybrid (BEH) technology and diol-bonded surface coating. Ideal for separating proteins from 10,000 to 450,000 Daltons, it offers mobile-phase pH stability for extended column lifetime. Suitable for quality control or LC-MS assays, it ensures durability, assay robustness, and exceptional efficiency.
  • Utilizes ethylene bridged hybrid (BEH)-based particle technology with diol-bonded surface coating.
  • Separates proteins within a 10,000 to 450,000 Dalton molecular weight range.
  • Ensures mobile-phase pH stability, extending column lifetime.
  • Supports quality control procedures and LC-MS assays.
  • Delivers column durability, assay robustness, and exceptional efficiency.
  • Includes one vial of protein standard at no additional cost.