Learn More
Waters Corp Waters Guard Column, 450Å, 3.5 μm, 7.8 mm X 30 mm
Supplier:  Waters Corp 186007641
Encompass_Preferred
The XBridge Protein BEH SEC Guard Column uses ethylene bridged hybrid (BEH)-based particle technology and diol-bonded surface coating. It is optimized for separating proteins from 100K to 1,500K daltons, providing a cost-effective way to protect analytical columns.
  • Built upon ethylene bridged hybrid (BEH)-based particle technology with diol-bonded surface coating.
  • Optimized for separating proteins ranging from 100K to 1,500K daltons.
  • Ensures improved column stability, performance, and longevity through BEH base particle technology and diol bonding.
  • Offers cost-effective protection for analytical columns, utilizing high-performance packing materials.
  • Manufactured in cGMP, ISO9001, and ISO13485 certified facilities, ensuring reliability, reproducibility, and high quality.
  • Thoroughly tested for unparalleled longevity across a wide range of mobile phase pH and temperatures.
Catalog No. 50-203-8134