Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Guard Column, SEC, 125Å, 2.5 μm, 4.6 mm X 30 mm

Catalog No. 502038927 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This guard column utilizes 2.5 μm Ethylene Bridged Hybrid (BEH) particles with a diol-bonded surface, designed for separating proteins from 1,000 to 80,000 Daltons.

  • Particle size: 2.5 μm
  • Pore size: 125 Å
  • Dimensions: 4.6 mm X 30 mm
  • Chemistry: SEC
  • Separation mode: Gel Filtration (aqueous)
  • Particle substrate: Hybrid
  • pH range: 2.5 to 8 pH
  • Temperature limits: 60 °C
  • Maximum pressure: 5500 psi (379 Bar)
  • Bonding technology: Diol (OH)
  • Silanol activity: Low
  • Molecular weight range: 1,000 to 80,000 Daltons
  • Particle shape: Spherical
  • Endfitting type: Parker-style
  • QC tested: Protein
  • Packing solvent: 20% methanol in water
  • Surface area: 220
  • System: UHPLC, UPLC, HPLC
  • USP classification: L20, L33, L59
  • Carbon load: 12 %

Catalog No. 50-203-8927 Supplier Waters Corp Supplier No. 186009170
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This guard column utilizes 2.5 μm Ethylene Bridged Hybrid (BEH) particles with a diol-bonded surface, designed for separating proteins from 1,000 to 80,000 Daltons.

  • Particle size: 2.5 μm
  • Pore size: 125 Å
  • Dimensions: 4.6 mm X 30 mm
  • Chemistry: SEC
  • Separation mode: Gel Filtration (aqueous)
  • Particle substrate: Hybrid
  • pH range: 2.5 to 8 pH
  • Temperature limits: 60 °C
  • Maximum pressure: 5500 psi (379 Bar)
  • Bonding technology: Diol (OH)
  • Silanol activity: Low
  • Molecular weight range: 1,000 to 80,000 Daltons
  • Particle shape: Spherical
  • Endfitting type: Parker-style
  • QC tested: Protein
  • Packing solvent: 20% methanol in water
  • Surface area: 220
  • System: UHPLC, UPLC, HPLC
  • USP classification: L20, L33, L59
  • Carbon load: 12 %

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.