Learn More
Waters Corp XBridge Protein BEH SEC Guard Column, 200Å, 2.5 μm, 4.6 mm X 30 mm
Catalog No. 502390453 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Protein BEH SEC Guard Column is designed for Size Exclusion Chromatography (SEC) applications, utilizing 2.5 μm Ethylene Bridged Hybrid (BEH)-based particles with a diol-bonded surface. It separates proteins from 10,000 to 450,000 Daltons and is compatible with UHPLC, UPLC, and HPLC systems for gel filtration in aqueous systems.
  • Utilizes ethylene bridged hybrid (BEH)-based particle technology for robust and reliable separations.
  • Optimized diol-bonded surface chemistry for consistent performance.
  • Separates proteins from 10,000 to 450,000 Daltons.
  • Compatible with UHPLC, UPLC, and HPLC systems.
  • Protects analytical columns by trapping particulates and strongly retained compounds.
  • Features low silanol activity for minimal secondary interactions.
  • Withstands pressures up to 4500 psi.
  • Designed for gel filtration in aqueous environments.
Catalog No. 50-239-0453 Supplier Waters Corp Supplier No. 186009174
The XBridge Protein BEH SEC Guard Column is designed for Size Exclusion Chromatography (SEC) applications, utilizing 2.5 μm Ethylene Bridged Hybrid (BEH)-based particles with a diol-bonded surface. It separates proteins from 10,000 to 450,000 Daltons and is compatible with UHPLC, UPLC, and HPLC systems for gel filtration in aqueous systems.
  • Utilizes ethylene bridged hybrid (BEH)-based particle technology for robust and reliable separations.
  • Optimized diol-bonded surface chemistry for consistent performance.
  • Separates proteins from 10,000 to 450,000 Daltons.
  • Compatible with UHPLC, UPLC, and HPLC systems.
  • Protects analytical columns by trapping particulates and strongly retained compounds.
  • Features low silanol activity for minimal secondary interactions.
  • Withstands pressures up to 4500 psi.
  • Designed for gel filtration in aqueous environments.