Learn More
Waters Corp Waters Corporation SEC Column, 200Å, 3.5 μm, 7.8 mm, 150 mm
Catalog No. 501015503 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These SEC columns, featuring ethylene bridged hybrid (BEH)-based particle technology and a diol-bonded surface coating, are designed for separating proteins ranging from 10,000 to 450,000 Daltons. They ensure reduced non-desired protein interactions and extended column life.
  • Utilizes ethylene bridged hybrid (BEH) particle technology with a diol-bonded surface coating.
  • Designed for efficient separation of proteins ranging from 10,000 to 450,000 Daltons.
  • Minimizes non-desired protein interactions, contributing to exceptional column life.
  • Provides flow and pressure tolerance for increased sample throughput on HPLC systems.
  • Delivers enhanced column stability, performance, and lifetime.
  • Includes a protein standard mix for comprehensive system benchmarking.
  • Manufactured in cGMP, ISO9001, and ISO13485 certified facilities ensuring quality.
Catalog No. 50-101-5503 Supplier Waters Corp Supplier No. 176003595
These SEC columns, featuring ethylene bridged hybrid (BEH)-based particle technology and a diol-bonded surface coating, are designed for separating proteins ranging from 10,000 to 450,000 Daltons. They ensure reduced non-desired protein interactions and extended column life.
  • Utilizes ethylene bridged hybrid (BEH) particle technology with a diol-bonded surface coating.
  • Designed for efficient separation of proteins ranging from 10,000 to 450,000 Daltons.
  • Minimizes non-desired protein interactions, contributing to exceptional column life.
  • Provides flow and pressure tolerance for increased sample throughput on HPLC systems.
  • Delivers enhanced column stability, performance, and lifetime.
  • Includes a protein standard mix for comprehensive system benchmarking.
  • Manufactured in cGMP, ISO9001, and ISO13485 certified facilities ensuring quality.