Learn More
Waters Corp Waters XBridge BEH Shield RP18 Column, 130Å, 3.5 μm, 4.6 mm x 75 mm
Catalog No. 50781496 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Shield RP18 columns provide alternative selectivity for phenolic compounds, featuring an embedded carbamate group. Its “Shield Technology” reduces silanol interactions for symmetrical and efficient peaks. This design enhances particle efficiency, peak form, and capacity, with improved pH stability and longer column life. All columns are rigorously tested for reproducibility and quality.
  • Complements C18 and C8 phases.
  • Offers alternative selectivity for phenolic compounds.
  • “Shield Technology” improves peak symmetry and efficiency.
  • Enhances particle efficiency and pH stability.
  • Rigorously tested for reproducibility.
  • Operates within 2–11 pH and up to 6000 psi (415 Bar).
Catalog No. 50-781-496 Supplier Waters Corp Supplier No. 186003043
XBridge BEH Shield RP18 columns provide alternative selectivity for phenolic compounds, featuring an embedded carbamate group. Its “Shield Technology” reduces silanol interactions for symmetrical and efficient peaks. This design enhances particle efficiency, peak form, and capacity, with improved pH stability and longer column life. All columns are rigorously tested for reproducibility and quality.
  • Complements C18 and C8 phases.
  • Offers alternative selectivity for phenolic compounds.
  • “Shield Technology” improves peak symmetry and efficiency.
  • Enhances particle efficiency and pH stability.
  • Rigorously tested for reproducibility.
  • Operates within 2–11 pH and up to 6000 psi (415 Bar).