Learn More
Waters Corp Waters HPLC Column, Shield RP18, 3.5 μm, 2.1 mm X 100 mm
Supplier:  Waters Corp 186003037
Encompass_Preferred
This XBridge BEH Shield RP18 Column complements C18 and C8 phases for HPLC method development. Its embedded carbamate group provides alternate selectivity for phenolic compounds. Engineered for chemical and mechanical stability, it withstands extreme backpressures and chromatographic conditions.
  • Provides unique selectivity for phenolic compounds.
  • MaxPeak HPS technology helps prevent unwanted analyte-surface interactions.
  • Features an embedded carbamate group in the bonded phase ligand for alternate selectivity.
  • Complements XBridge BEH C18 and C8 phases.
  • Ethylene-Bridged Hybrid (BEH) base particle ensures maximum column performance and elongated column lifetimes.
  • Offers improved basic analyte peak shape near neutral pH.
  • Can accommodate 100% aqueous mobile phases.
  • Can be protected from contaminants with the XBridge BEH Shield RP18 VanGuard Cartridge.
Catalog No. 50-781-490