Learn More
Waters Corp Waters HPLC Column, 130Å, 3.5μm, 2.1 mm × 30 mm
Catalog No. 50781488 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Shield RP18 columns complement the XBridge BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.
  • Complement BEH C18 and C8 phases
  • Powerful tool for HPLC method development
  • Embedded carbamate group in the bonded phase ligand provides alternate selectivity
  • Designed for phenolic compounds
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 2 to 11
  • Maximum pressure of 6000 psi (415 bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • BEH particle technology
  • L1 USP classification
  • 17% carbon load
Catalog No. 50-781-488 Supplier Waters Corp Supplier No. 186003035
XBridge BEH Shield RP18 columns complement the XBridge BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.
  • Complement BEH C18 and C8 phases
  • Powerful tool for HPLC method development
  • Embedded carbamate group in the bonded phase ligand provides alternate selectivity
  • Designed for phenolic compounds
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range of 2 to 11
  • Maximum pressure of 6000 psi (415 bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • BEH particle technology
  • L1 USP classification
  • 17% carbon load