Learn More
Waters Corp Waters XBridge BEH Shield RP18 Column, 130Å, 3.5 μm, 3 mm X 100 mm
Catalog No. 50781493 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Shield RP18 columns are engineered to complement BEH C18 and C8 phases, providing an alternative selectivity for HPLC method development, especially for phenolic compounds. They minimize silanol interactions with basic analytes through a “Shielding” technique, ensuring symmetrical and efficient peaks.
  • Offers alternative selectivity, particularly for phenolic compounds.
  • Reduces silanol interactions with basic analytes.
  • Provides exceptionally symmetrical, efficient peaks with superior peak shapes.
  • Suitable for use with various solvents and across a wide pH range.
  • Offers exceptional pH stability, high efficiency, and symmetrical peak form.
  • Enables seamless UPLC transferability.
  • Suitable for preparative scaling for product isolation.
  • Recommended maximum pressure of 6000 psi (415 Bar).
Catalog No. 50-781-493 Supplier Waters Corp Supplier No. 186003040
XBridge BEH Shield RP18 columns are engineered to complement BEH C18 and C8 phases, providing an alternative selectivity for HPLC method development, especially for phenolic compounds. They minimize silanol interactions with basic analytes through a “Shielding” technique, ensuring symmetrical and efficient peaks.
  • Offers alternative selectivity, particularly for phenolic compounds.
  • Reduces silanol interactions with basic analytes.
  • Provides exceptionally symmetrical, efficient peaks with superior peak shapes.
  • Suitable for use with various solvents and across a wide pH range.
  • Offers exceptional pH stability, high efficiency, and symmetrical peak form.
  • Enables seamless UPLC transferability.
  • Suitable for preparative scaling for product isolation.
  • Recommended maximum pressure of 6000 psi (415 Bar).