Learn More
Waters Corp Waters Corporation Column, XBridge BEH Shield RP18, 130Å, 3.5 μm
Catalog No. 50781498 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These BEH Shield RP18 columns are designed for high-performance liquid chromatography (HPLC) method development, offering alternate selectivity for compounds like phenolics. They deliver excellent peak shape, high efficiency, and stability across acidic and basic mobile phases, enabling robust and confident separations through advanced bonding and particle technology.
  • Complements C18/C8 phases for HPLC method development with alternate selectivity.
  • Embedded carbamate group provides specific selectivity for phenolic compounds.
  • Achieves excellent peak shape, high efficiency, and stability in acidic/basic mobile phases.
  • Shield Technology ensures stable retention in 100% aqueous mobile phases.
  • BEH particle offers wide usable pH range (pH 1–12) with superior low pH stability.
Catalog No. 50-781-498 Supplier Waters Corp Supplier No. 186003045
Add to Cart
Add to Cart
These BEH Shield RP18 columns are designed for high-performance liquid chromatography (HPLC) method development, offering alternate selectivity for compounds like phenolics. They deliver excellent peak shape, high efficiency, and stability across acidic and basic mobile phases, enabling robust and confident separations through advanced bonding and particle technology.
  • Complements C18/C8 phases for HPLC method development with alternate selectivity.
  • Embedded carbamate group provides specific selectivity for phenolic compounds.
  • Achieves excellent peak shape, high efficiency, and stability in acidic/basic mobile phases.
  • Shield Technology ensures stable retention in 100% aqueous mobile phases.
  • BEH particle offers wide usable pH range (pH 1–12) with superior low pH stability.