Learn More
Waters Corp XBridge BEH Shield RP18 Column, 130Å, 3.5 μm, 4.6 mm, 100 mm
Catalog No. 50781497 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are designed to complement BEH C18 and C8 phases, providing an essential tool for HPLC method development. They feature an embedded carbamate group in the bonded phase ligand, which offers alternate selectivity, particularly for phenolic compounds, compared to traditional straight-chain alkyl columns.
  • Alternate selectivity for phenolic compounds with embedded carbamate group.
  • Improved peak shape by reducing silanol interactions with basic analytes.
  • Robust separations through full utilization of pH.
  • Superior pH stability over widest range (1 to 12) for method ruggedness.
  • Enhanced productivity, maximizing particle efficiency and column lifetime.
  • Unmatched reproducibility through controlled manufacturing and testing.
Catalog No. 50-781-497 Supplier Waters Corp Supplier No. 186003044
These columns are designed to complement BEH C18 and C8 phases, providing an essential tool for HPLC method development. They feature an embedded carbamate group in the bonded phase ligand, which offers alternate selectivity, particularly for phenolic compounds, compared to traditional straight-chain alkyl columns.
  • Alternate selectivity for phenolic compounds with embedded carbamate group.
  • Improved peak shape by reducing silanol interactions with basic analytes.
  • Robust separations through full utilization of pH.
  • Superior pH stability over widest range (1 to 12) for method ruggedness.
  • Enhanced productivity, maximizing particle efficiency and column lifetime.
  • Unmatched reproducibility through controlled manufacturing and testing.