Learn More
Waters Corp Waters Corporation XBridge BEH C18 OBD Prep Column, 10 x 250 mm, 5 μm
Catalog No. 50781581 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Corporation XBridge BEH C18 OBD Prep Column is designed for high-performance preparative chromatography. It features an Optimum Bed Density (OBD) design, enabling direct scalability from analytical to preparative scale while maintaining maximum efficiency and long column lifetimes.
  • Optimum bed density (OBD) design
  • Direct scalability from analytical to preparative scale
  • Maximum efficiency
  • Long column lifetimes
  • Lower column backpressure
  • High loading capacity
  • Exceptional resistance to mechanical chromatographic bed failure
  • Consistent column-to-column performance
  • Reduced cost through extended lifetimes
Catalog No. 50-781-581 Supplier Waters Corp Supplier No. 186003149
The Waters Corporation XBridge BEH C18 OBD Prep Column is designed for high-performance preparative chromatography. It features an Optimum Bed Density (OBD) design, enabling direct scalability from analytical to preparative scale while maintaining maximum efficiency and long column lifetimes.
  • Optimum bed density (OBD) design
  • Direct scalability from analytical to preparative scale
  • Maximum efficiency
  • Long column lifetimes
  • Lower column backpressure
  • High loading capacity
  • Exceptional resistance to mechanical chromatographic bed failure
  • Consistent column-to-column performance
  • Reduced cost through extended lifetimes