Learn More
Waters Corp Waters Column, XBridge BEH Shield RP18, 130Å, 5 μm, 4.6 mm X 75 mm
Catalog No. 50781460 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Shield RP18 columns are designed for HPLC method development, offering alternate selectivity for compounds like phenolics. They incorporate MaxPeak High-Performance Surface technology to prevent unwanted analyte-surface interactions. These columns provide a wide and stable pH range (2 to 11) and utilize Ethylene-Bridged Hybrid (BEH) particle technology for mechanical strength and extended lifetime.
  • Alternate selectivity, especially for phenolic compounds
  • MaxPeak High-Performance Surface technology prevents unwanted analyte-surface interactions
  • Wide and usable pH range (2 to 11) with chemical stability
  • Ethylene-Bridged Hybrid (BEH) particle technology ensures mechanical strength at high backpressures
  • Improved basic analyte peak shape near neutral pH
  • Extended column lifetime
  • Protection from particulate contaminants with a compatible VanGuard Cartridge
Catalog No. 50-781-460 Supplier Waters Corp Supplier No. 186003007
XBridge BEH Shield RP18 columns are designed for HPLC method development, offering alternate selectivity for compounds like phenolics. They incorporate MaxPeak High-Performance Surface technology to prevent unwanted analyte-surface interactions. These columns provide a wide and stable pH range (2 to 11) and utilize Ethylene-Bridged Hybrid (BEH) particle technology for mechanical strength and extended lifetime.
  • Alternate selectivity, especially for phenolic compounds
  • MaxPeak High-Performance Surface technology prevents unwanted analyte-surface interactions
  • Wide and usable pH range (2 to 11) with chemical stability
  • Ethylene-Bridged Hybrid (BEH) particle technology ensures mechanical strength at high backpressures
  • Improved basic analyte peak shape near neutral pH
  • Extended column lifetime
  • Protection from particulate contaminants with a compatible VanGuard Cartridge