Learn More
Waters Corp XBridge Column, 130Å, 5 μm, 4.6 mm × 250 mm
Supplier:  Waters Corp 186003010
Encompass_Preferred
These BEH Shield RP18 columns are designed for HPLC method development, complementing C18 and C8 phases. The embedded carbamate group provides alternate selectivity, especially for phenolic compounds, and allows operation across a wide pH range with various solvents.
  • Optimize test conditions for successful separation.
  • Offer alternate selectivity by operating over a wide pH range with various solvents.
  • Achieve superior separation of bases with reduced silanol interactions.
  • Patented Shield Technology leads to highly symmetrical and efficient peaks.
  • This “Shielding” effect improves peak shape.
  • Enable robust separations with less time and greater confidence, fully utilizing pH.
Catalog No. 50-781-463