Learn More
Waters Corp Waters XBridge BEH Shield RP18 Column, 130Å, 5 μm, 4.6 × 50 mm
Supplier:  Waters Corp 186003006
Encompass_Preferred
XBridge BEH Shield RP18 columns are essential for HPLC method development, offering alternative selectivity for phenolic compounds with their unique bonded phase ligand. Manufactured in state-of-the-art facilities, these columns ensure batch-to-batch reproducibility, stability, and high quality through rigorous testing and adherence to industry standards.
  • Offers alternate selectivity for phenolic compounds.
  • Enhances particle efficiency.
  • Achieves unrivaled peak shape and capacity.
  • Provides high accuracy and efficiency.
  • Improves pH stability for greater column lifetime.
  • Aids in improved base separation.
  • Reduces silanol interactions for symmetrical peaks.
  • Ensures repeatable column performance and batch-to-batch reproducibility.
  • Extensively tested for highest quality.
Catalog No. 50-781-459