Learn More
Waters Corp Waters Column, Shield RP18, 5 μm, 4.6 mm X 100 mm, 130 Å
Catalog No. 50781461 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Shield RP18 columns offer alternate selectivity, especially for phenolic compounds, due to an embedded carbamate group. This design reduces silanol interactions with basic analytes, leading to highly symmetrical peaks and improved pH stability for increased column lifetime and assay ruggedness.
  • Embedded carbamate group provides alternate selectivity for phenolic compounds.
  • Shield Technology reduces silanol interactions with basic analytes, improving peak shape.
  • Improved pH stability increases column lifetime and assay ruggedness.
  • Manufactured under controlled processes for batch-to-batch reproducibility.
  • Ensures stable retention in 100% aqueous mobile phases, preventing dewetting.
  • Maximizes particle efficiency for unmatched peak shape and capacity.
Catalog No. 50-781-461 Supplier Waters Corp Supplier No. 186003008
XBridge BEH Shield RP18 columns offer alternate selectivity, especially for phenolic compounds, due to an embedded carbamate group. This design reduces silanol interactions with basic analytes, leading to highly symmetrical peaks and improved pH stability for increased column lifetime and assay ruggedness.
  • Embedded carbamate group provides alternate selectivity for phenolic compounds.
  • Shield Technology reduces silanol interactions with basic analytes, improving peak shape.
  • Improved pH stability increases column lifetime and assay ruggedness.
  • Manufactured under controlled processes for batch-to-batch reproducibility.
  • Ensures stable retention in 100% aqueous mobile phases, preventing dewetting.
  • Maximizes particle efficiency for unmatched peak shape and capacity.