Learn More
Waters Corp XBridge Shield RP18 Column, 5 μm, 3 mm x 50 mm
Catalog No. 50781592 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Shield RP18 HPLC Column is engineered to provide complementary selectivity and superior peak shape for basic compounds, making it an ideal choice for methods development scientists seeking alternative selectivity to traditional C18 chemistries.
  • Patented Shield Technology for improved peak shapes.
  • BEH (Ethylene Bridged Hybrid) particle technology for stability and extended column lifetime.
  • 5 μm particle size.
  • Column configuration: 3 mm x 50 mm.
  • Operates over a broad pH range of 2 to 11.
  • Aqueous compatibility without risk of pore dewetting.
  • Nominal pore diameter: 135 Å.
  • Superior separation of basic compounds.
  • Unique selectivity for method development.
  • Robust performance and longevity.
Catalog No. 50-781-592 Supplier Waters Corp Supplier No. 186003160
Add to Cart
Add to Cart
The XBridge Shield RP18 HPLC Column is engineered to provide complementary selectivity and superior peak shape for basic compounds, making it an ideal choice for methods development scientists seeking alternative selectivity to traditional C18 chemistries.
  • Patented Shield Technology for improved peak shapes.
  • BEH (Ethylene Bridged Hybrid) particle technology for stability and extended column lifetime.
  • 5 μm particle size.
  • Column configuration: 3 mm x 50 mm.
  • Operates over a broad pH range of 2 to 11.
  • Aqueous compatibility without risk of pore dewetting.
  • Nominal pore diameter: 135 Å.
  • Superior separation of basic compounds.
  • Unique selectivity for method development.
  • Robust performance and longevity.